Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF013249.1 | - | 3 | 259 | ||
AF109683.1 | - | 3 | 275 | |||
AF251509.2 | - | 3 | 191 | |||
AF251510.2 | - | 3 | 194 | |||
AK097869.1 | - | 2 | 382 | |||
AK290415.1 | - | 3 | 315 | |||
AK303261.1 | - | 3 | 345 | |||
AK309895.1 | - | 3 | 271 | |||
AK315419.1 | - | 3 | 347 | |||
BC027899.1 | - | 3 | 251 | 5228732 | ||
BI756388.1 | - | 3 | 257 | |||
BM918573.1 | - | 3 | 379 | |||
CR542051.1 | - | 3 | 191 | |||
DA419411.1 | - | 3 | 224 | |||
DB143367.1 | - | 2 | 414 | |||
DC379147.1 | - | 3 | 478 | |||
KU058457.1 | - | 446 |