Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284328.1 | NP_001271257.1 | 9 | 1455 | ||
NM_019666.2 | NP_062640.2 | 9 | 1455 | |||
NM_019796.5 | NP_062770.1 | 9 | 1455 | |||
GenBank mRNA | AB035725.1 | - | 9 | 1254 | ||
AF093821.1 | - | 8 | 1116 | |||
AF408434.1 | - | 7 | 908 | |||
AK034845.1 | - | 9 | 1267 | |||
AK076026.1 | - | 9 | 1245 | |||
AK077588.1 | - | 9 | 1204 | |||
AK078158.1 | - | 7 | 888 | |||
AK083398.1 | - | 8 | 1046 | |||
AK162448.1 | - | 3 | 337 | |||
BC041148.1 | - | 9 | 1300 | 4946652 | ||
BC050079.1 | - | 9 | 1228 | 30012104 | ||
BC055863.1 | - | 9 | 1233 | |||
BC058807.1 | - | 9 | 1322 | |||
BC080309.1 | - | 9 | 1158 | 5716184 | ||
BC108363.1 | - | 9 | 1189 | 4239809 |