Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019819.3 | NP_062793.2 | 3 | 336 | ||
GenBank mRNA | AF120113.1 | - | 1 | 62 | ||
AK009744.1 | - | 3 | 349 | |||
AK079362.1 | - | 3 | 337 | |||
AK163738.1 | - | 3 | 381 | |||
AK166230.1 | - | 3 | 334 | |||
AK166840.1 | - | 3 | 332 | |||
BC002130.1 | - | 3 | 301 | 3592420 | ||
BC052836.1 | - | 3 | 356 | 30049007 |