Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291071.1 | NP_001278000.1 | 20 | 2130 | ||
NM_019703.4 | NP_062677.1 | 20 | 2130 | |||
GenBank mRNA | AF123533.1 | - | 20 | 2095 | ||
AK004790.1 | - | 20 | 2084 | |||
AK047303.1 | - | 2 | 129 | |||
AK049841.1 | - | 20 | 2084 | |||
AK076803.1 | - | 20 | 2085 | |||
AK078254.1 | - | 20 | 2085 | |||
AK098162.1 | - | 20 | 2084 | |||
AK170624.1 | - | 20 | 2085 | |||
AY779276.1 | - | 20 | 2047 | |||
BC006926.1 | - | 20 | 2069 | 2655383 | ||
CT010268.1 | - | 20 | 2038 | |||
Y19008.1 | - | 20 | 2087 |