Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019771.2 | NP_062745.1 | 2 | 550 | ||
GenBank mRNA | AB025406.1 | - | 2 | 270 | ||
AK078898.1 | - | 2 | 324 | |||
BC131926.1 | - | 2 | 267 | 40130203 | ||
BF123977.1 | - | 2 | 561 | |||
CT010216.1 | - | 1 | 236 |