Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019712.3 | NP_062686.1 | 3 | 252 | ||
GenBank mRNA | AF140599.1 | - | 3 | 212 | ||
AK003159.1 | - | 3 | 214 | |||
AK004114.1 | - | 3 | 216 | |||
AK149259.1 | - | 2 | 1455 | |||
AK152012.1 | - | 3 | 209 | |||
AK159571.1 | - | 3 | 209 | |||
AK182752.1 | - | 3 | 146 | |||
AK183716.1 | - | 3 | 146 | |||
BC027396.1 | - | 3 | 201 | 4952242 | ||
BC051473.1 | - | 3 | 207 | 1430400 | ||
BC056992.1 | - | 3 | 195 | 6818827 | ||
CJ082632.1 | - | 3 | 272 |