Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019727.2 | NP_062701.2 | 8 | 887 | ||
GenBank mRNA | AB019214.1 | - | 8 | 827 | ||
AF154120.1 | - | 8 | 891 | |||
AK149850.1 | - | 8 | 799 | |||
AK154079.1 | - | 8 | 800 | |||
AK155746.1 | - | 8 | 775 | |||
AK161241.1 | - | 8 | 868 | |||
AK167177.1 | - | 8 | 842 | |||
AK167993.1 | - | 8 | 782 | |||
AK168742.1 | - | 8 | 880 | |||
AK177911.1 | - | 2 | 77 | |||
AK192993.1 | - | 2 | 77 | |||
AK193915.1 | - | 2 | 77 | |||
BC066044.1 | - | 8 | 844 | 6807363 | ||
BC066189.1 | - | 8 | 778 | 30471747 |