Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019727.2 | NP_062701.2 | 12 | 1470 | ||
GenBank mRNA | AB019214.1 | - | 12 | 1410 | ||
AF154120.1 | - | 12 | 1474 | |||
AK076198.1 | - | 4 | 454 | |||
AK149850.1 | - | 12 | 1382 | |||
AK154079.1 | - | 12 | 1383 | |||
AK155746.1 | - | 12 | 1358 | |||
AK161241.1 | - | 12 | 1451 | |||
AK167177.1 | - | 12 | 1425 | |||
AK167993.1 | - | 12 | 1365 | |||
AK168742.1 | - | 12 | 1463 | |||
AK189858.1 | - | 1 | 62 | |||
AK197557.1 | - | 1 | 62 | |||
AK201929.1 | - | 1 | 62 | |||
AK202656.1 | - | 1 | 62 | |||
AK203711.1 | - | 1 | 62 | |||
BC066044.1 | - | 12 | 1427 | 6807363 | ||
BC066189.1 | - | 12 | 1361 | 30471747 |