Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019785.2 | NP_062759.2 | 3 | 262 | ||
GenBank mRNA | AF190797.1 | - | 3 | 229 | ||
AK084255.1 | - | 1 | 1929 | |||
AK090043.1 | - | 3 | 266 | |||
AK161642.1 | - | 3 | 222 | |||
BC002227.1 | - | 3 | 222 | 3491299 |