Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 24, 25 | 3244 | ||
NM_001127717.2 | NP_001121189.2 | 60, 61 | 8119 | |||
NM_001127718.2 | NP_001121190.2 | 23, 24 | 3076 | |||
NM_198129.2 | NP_937762.2 | 61, 62 | 8287 | |||
XM_011525978.2 | XP_011524280.1 | 61, 62 | 8314 | |||
XM_011525979.2 | XP_011524281.1 | 61, 62 | 8305 | |||
XM_011525980.2 | XP_011524282.1 | 61, 62 | 8296 | |||
XM_011525981.2 | XP_011524283.1 | 60, 61 | 8182 | |||
XM_011525982.2 | XP_011524284.1 | 59, 60 | 8017 | |||
XM_017025743.1 | XP_016881232.1 | 44, 45 | 5998 | |||
XM_017025744.1 | XP_016881233.1 | 34, 35 | 4679 | |||
GenBank mRNA | AB107369.1 | - | 61, 62 | 8046 | ||
AB208853.1 | - | 16, 17 | 2272 | |||
AY327114.1 | - | 23, 24 | 3051 | |||
AY327115.1 | - | 61, 62 | 8046 | |||
AY327116.1 | - | 60, 61 | 7878 | |||
BC043618.1 | - | 18, 19 | 2624 | |||
L34155.1 | - | 24, 25 | 3186 | |||
X84900.1 | - | 26, 27 | 3465 | |||
X85107.1 | - | 24, 25 | 3237 | |||
X85108.1 | - | 26, 27 | 3465 |