Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285891.1 | NP_001272820.1 | 4, 5 | 678 | ||
NM_001285892.1 | NP_001272821.1 | 4, 5 | 678 | |||
NM_019748.3 | NP_062722.1 | 4, 5 | 678 | |||
GenBank mRNA | AB024303.1 | - | 4, 5 | 566 | ||
AK010313.1 | - | 4, 5 | 555 | |||
AK011772.1 | - | 4, 5 | 589 | |||
AK011783.1 | - | 1, 2 | 25 | |||
AK087556.1 | - | 4, 5 | 569 | |||
AK090012.1 | - | 4, 5 | 565 | |||
AK154139.1 | - | 4, 5 | 591 | |||
AK159672.1 | - | 4, 5 | 580 | |||
AK162789.1 | - | 4, 5 | 566 | |||
BC068164.1 | - | 4, 5 | 567 | 30359189 |