Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291205.1 | NP_001278134.1 | 5 | 544 | ||
NM_001291206.1 | NP_001278135.1 | 5 | 895 | |||
NM_001291207.1 | NP_001278136.1 | 4 | 440 | |||
NM_001291208.1 | NP_001278137.1 | 4 | 789 | |||
NM_001291209.1 | NP_001278138.1 | 5 | 546 | |||
NM_001291210.1 | NP_001278139.1 | 3 | 318 | |||
NM_026446.4 | NP_080722.1 | 5 | 546 | |||
GenBank mRNA | AF535869.1 | - | 5 | 896 | ||
AF535870.1 | - | 4 | 790 | |||
AK011740.2 | - | 5 | 495 | |||
AK019401.1 | - | 5 | 519 | |||
AK042531.1 | - | 5 | 494 | |||
AK154844.1 | - | 5 | 546 | |||
AY138505.1 | - | 4 | 392 | |||
BC003838.1 | - | 5 | 478 | 3490927 | ||
BU694737.1 | - | 3 | 354 | |||
CF751080.1 | - | 4 | 460 |