Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019699.1 | NP_062673.1 | 8 | 993 | ||
GenBank mRNA | AF126798.1 | - | 8 | 993 | ||
AK083282.1 | - | 960 | ||||
AK143524.1 | - | 1011 | ||||
BC057189.1 | - | 8 | 981 | 5137343 | ||
HQ264057.1 | - | 8 | 886 | |||
HQ264058.1 | - | 8 | 886 |