Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005560.4 | NP_005551.3 | 61 | 8400 | ||
XM_006723796.3 | XP_006723859.1 | 61 | 8403 | |||
XM_011528818.2 | XP_011527120.1 | 60 | 8265 | |||
XM_011528819.2 | XP_011527121.1 | 59 | 8130 | |||
GenBank mRNA | AB011105.1 | - | 16 | 2185 | ||
AF443072.1 | - | 61 | 8400 | |||
Z95636.1 | - | 1 | 107 |