Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019656.3 | NP_062630.2 | 5 | 605 | ||
GenBank mRNA | AF053454.1 | - | 5 | 552 | ||
AK146673.1 | - | 5 | 510 | |||
AK149204.1 | - | 5 | 515 | |||
AK160313.1 | - | 5 | 499 | |||
AK167384.1 | - | 5 | 501 | |||
BC003733.1 | - | 5 | 539 | 3590862 | ||
BC110314.1 | - | 5 | 514 | 5344878 |