Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019680.2 | NP_062654.2 | 9 | 1586 | ||
GenBank mRNA | AF016714.1 | - | 8 | 1527 | ||
AK155203.1 | - | 9 | 1827 | |||
BC109160.1 | - | 8 | 1518 | 40047677 | ||
BC109171.1 | - | 8 | 1518 | 40048316 |