Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019686.5 | NP_062660.1 | 1, 2 | 121 | ||
GenBank mRNA | AB016080.1 | - | 1, 2 | 121 | ||
AK005347.1 | - | 1, 2 | 225 | |||
AK013240.1 | - | 230 | ||||
BC005739.1 | - | 1, 2 | 163 | 3599342 |