Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 818 | ||
NM_001047436.2 | NP_001040901.1 | 3 | 834 | |||
NM_001286344.1 | NP_001273273.1 | 3 | 803 | |||
NM_001286345.1 | NP_001273274.1 | 3 | 731 | |||
NM_019695.3 | NP_062669.2 | 3 | 821 | |||
GenBank mRNA | AF070970.1 | - | 3 | 817 | ||
AK003030.1 | - | 3 | 787 | |||
AK158848.1 | - | 3 | 816 | |||
AK209727.1 | - | 1 | 56 | |||
BC049593.1 | - | 3 | 795 | 6771847 | ||
BC061114.1 | - | 3 | 731 | 6535523 |