Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 849 | ||
NM_001047436.2 | NP_001040901.1 | 3 | 865 | |||
NM_001286344.1 | NP_001273273.1 | 3 | 834 | |||
NM_001286345.1 | NP_001273274.1 | 3 | 762 | |||
NM_019695.3 | NP_062669.2 | 3 | 852 | |||
GenBank mRNA | AF070970.1 | - | 3 | 848 | ||
AK003030.1 | - | 3 | 818 | |||
AK158848.1 | - | 3 | 847 | |||
AK209727.1 | - | 1 | 87 | |||
BC049593.1 | - | 3 | 826 | 6771847 | ||
BC061114.1 | - | 3 | 762 | 6535523 |