Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019723.2 | NP_062697.1 | 4 | 847 | ||
GenBank mRNA | AB018436.1 | - | 4 | 847 | ||
AK080177.1 | - | 3 | 1050 | |||
AK133431.1 | - | 4 | 776 | |||
AK161508.1 | - | 4 | 743 | |||
BC132147.1 | - | 4 | 810 | 40130424 | ||
BC137908.1 | - | 4 | 810 | 8860927 |