Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019728.4 | NP_062702.1 | 2 | 120 | ||
GenBank mRNA | AF155882.2 | - | 2 | 99 | ||
AK009061.1 | - | 2 | 119 | |||
AK009306.1 | - | 2 | 119 | |||
BC099882.1 | - | 2 | 119 | 40040625 | ||
BC099883.1 | - | 2 | 119 | 40040627 | ||
BC100674.1 | - | 2 | 119 | 40040624 | ||
BC100675.1 | - | 2 | 119 | 40040630 | ||
EH111306.1 | - | 2 | 120 |