Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000228.2 | NP_000219.2 | 4 | 380 | ||
NM_001017402.1 | NP_001017402.1 | 3 | 626 | |||
NM_001127641.1 | NP_001121113.1 | 4 | 354 | |||
XM_005273124.4 | XP_005273181.1 | 4 | 562 | |||
XM_017001272.1 | XP_016856761.1 | 4 | 982 | |||
GenBank mRNA | AK296851.1 | - | 4 | 330 | ||
AK309524.1 | - | 4 | 330 | |||
AL555685.3 | - | 4 | 380 | |||
AY035783.1 | - | 3 | 236 | |||
BC075838.1 | - | 3 | 411 | 6291096 | ||
BP350980.1 | - | 4 | 396 | |||
BX345166.2 | - | 3 | 384 | |||
D37766.1 | - | 4 | 317 | |||
KU178159.1 | - | 3 | 236 | |||
L25541.1 | - | 4 | 356 |