Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177323.2 | NP_001170794.1 | 5 | 897 | ||
NM_001177324.1 | NP_001170795.1 | 5 | 900 | |||
NM_001253706.1 | NP_001240635.1 | 5 | 900 | |||
NM_019942.5 | NP_064326.2 | 5 | 900 | |||
GenBank mRNA | AB023622.1 | - | 5 | 664 | ||
AK012858.1 | - | 6 | 694 | |||
AK048455.1 | - | 5 | 722 | |||
AK088406.1 | - | 5 | 680 | |||
AK088614.1 | - | 5 | 663 | |||
AK160884.1 | - | 5 | 694 | |||
AK162755.1 | - | 5 | 700 | |||
AK172894.1 | - | 5 | 681 | |||
BC010489.1 | - | 5 | 686 | 4168214 |