Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019945.2 | NP_064329.2 | 26 | 4616 | ||
GenBank mRNA | AF077818.1 | - | 26 | 4590 | ||
AK122411.1 | - | 15 | 3825 | |||
BC039230.1 | - | 14 | 4568 | |||
BC054524.1 | - | 26 | 4616 | 6400705 | ||
BU701555.1 | - | 1 | 310 |