Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161822.1 | NP_001155294.1 | 5 | 587 | ||
NM_019958.4 | NP_064342.1 | 4 | 517 | |||
GenBank mRNA | AK018279.1 | - | 4 | 501 | ||
AK030492.1 | - | 4 | 518 | |||
AK045770.1 | - | 5 | 554 | |||
AK046009.1 | - | 4 | 476 | |||
AK078745.1 | - | 4 | 476 | |||
AK163188.1 | - | 4 | 474 | |||
BC064782.1 | - | 4 | 445 | 30476405 |