Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027044.3 | NP_081320.2 | 2, 3 | 249 | ||
GenBank mRNA | AF108357.1 | - | 2, 3 | 209 | ||
AK004438.1 | - | 2, 3 | 204 | |||
AK005682.1 | - | 2, 3 | 195 | |||
AK028206.1 | - | 2, 3 | 186 | |||
AK145883.1 | - | 2, 3 | 202 | |||
AK211903.1 | - | 1, 2 | 48 | |||
BC026920.1 | - | 2, 3 | 186 | 3666296 | ||
CN835522.1 | - | 2, 3 | 252 |