Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019924.2 | NP_064308.2 | 7, 8 | 842 | ||
GenBank mRNA | AF074714.1 | - | 7, 8 | 814 | ||
AK154674.1 | - | 7, 8 | 841 | |||
AK204246.1 | - | 2, 3 | 186 | |||
BC012964.1 | - | 7, 8 | 852 | 4160873 |