Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020032.2 | NP_064416.1 | 9 | 1616 | ||
GenBank mRNA | AF176099.1 | - | 8 | 1557 | ||
AJ131889.1 | - | 9 | 1616 | |||
AK003392.1 | - | 5 | 752 | |||
AK133087.1 | - | 9 | 1738 | |||
BC004767.1 | - | 8 | 1566 | 3156259 |