Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172561.1 | NP_001166032.1 | 6 | 1851 | ||
NM_020011.5 | NP_064395.2 | 6 | 2160 | |||
NM_203280.3 | NP_975009.1 | 7 | 1922 | |||
GenBank mRNA | AF245448.1 | - | 7 | 1872 | ||
AF415214.1 | - | 5 | 1486 | |||
AK004951.1 | - | 6 | 2160 | |||
AK016616.1 | - | 3 | 940 | |||
AK146414.1 | - | 6 | 1752 | |||
AK151990.1 | - | 7 | 1822 | |||
AK154321.1 | - | 6 | 1588 | |||
BC006941.1 | - | 6 | 1594 | 2650442 | ||
BC053737.1 | - | 7 | 1835 | 6507910 | ||
BC092084.1 | - | 6 | 1609 | 4987298 |