Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005562.2 | NP_005553.2 | 11, 12 | 2029 | ||
NM_018891.2 | NP_061486.2 | 11, 12 | 2029 | |||
XM_017001273.1 | XP_016856762.1 | 11, 12 | 1792 | |||
GenBank mRNA | AK296944.1 | - | 11, 12 | 1792 | ||
AK307945.1 | - | 11, 12 | 1842 | |||
BC112286.1 | - | 11, 12 | 1756 | 8327754 | ||
BC113378.1 | - | 11, 12 | 1756 | 8322430 | ||
BC143653.1 | - | 11, 12 | 1733 | |||
X73902.1 | - | 11, 12 | 1780 | |||
Z15008.1 | - | 11, 12 | 1832 | |||
Z15009.1 | - | 11, 12 | 1832 |