Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019928.1 | NP_064312.1 | 4 | 380 | ||
GenBank mRNA | AF019979.1 | - | 4 | 380 | ||
AK087426.1 | - | 4 | 394 | |||
BC100716.1 | - | 3 | 352 | 40045068 | ||
BC100717.1 | - | 3 | 352 | 40045067 | ||
BC100718.1 | - | 3 | 352 | 40045065 | ||
BC100719.1 | - | 3 | 352 | 40045063 |