Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053079.2 | NP_444309.2 | 18, 19 | 1509 | ||
GenBank mRNA | AF205540.1 | - | 18, 19 | 1508 | ||
AK136428.1 | - | 18, 19 | 1510 | |||
BC116248.1 | - | 18, 19 | 1499 | 40093541 | ||
BC116249.1 | - | 18, 19 | 1499 | 40093542 |