Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019920.2 | NP_064304.1 | 5 | 337 | ||
GenBank mRNA | AF082526.1 | - | 5 | 337 | ||
AK031303.1 | - | 1 | 723 | |||
AK088454.1 | - | 5 | 344 | |||
AK151432.1 | - | 5 | 367 | |||
AK159450.1 | - | 5 | 335 | |||
AK162102.1 | - | 5 | 329 | |||
BC016417.1 | - | 5 | 320 | 3158151 | ||
BC083155.1 | - | 5 | 319 | 5339040 |