Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 1, 2 | 268 | ||
GenBank mRNA | AK049399.1 | - | 1, 2 | 237 | ||
AK083255.1 | - | 1, 2 | 237 | |||
AK162966.1 | - | 1, 2 | 269 |