Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038560.1 | - | 1 | 897 | ||
AK050819.1 | - | 2 | 236 | |||
AK220315.1 | - | 1 | 845 | |||
BC036977.1 | - | 2 | 242 | |||
BC092394.1 | - | 1 | 915 | 30534262 | ||
BC141058.1 | - | 1 | 853 | 9055670 |