Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019937.3 | NP_064321.2 | 2 | 473 | ||
GenBank mRNA | AF159159.1 | - | 2 | 426 | ||
AF467251.1 | - | 2 | 467 | |||
AK011629.1 | - | 2 | 430 | |||
AK051380.1 | - | 2 | 430 | |||
BC007177.1 | - | 2 | 447 | |||
BC037204.1 | - | 2 | 411 | |||
BC069909.1 | - | 2 | 410 | |||
BC094383.1 | - | 2 | 409 | 30604933 | ||
BC117816.1 | - | 2 | 293 |