Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019937.3 | NP_064321.2 | 3 | 518 | ||
GenBank mRNA | AF159159.1 | - | 3 | 471 | ||
AF467251.1 | - | 3 | 512 | |||
AK011629.1 | - | 3 | 475 | |||
AK051380.1 | - | 3 | 475 | |||
BC007177.1 | - | 3 | 492 | |||
BC037204.1 | - | 3 | 456 | |||
BC069909.1 | - | 3 | 455 | |||
BC094383.1 | - | 3 | 454 | 30604933 | ||
BC117816.1 | - | 3 | 338 |