Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252463.1 | NP_001239392.1 | 4 | 396 | ||
NM_001252464.1 | NP_001239393.1 | 4 | 371 | |||
NM_001252465.1 | NP_001239394.1 | 2 | 180 | |||
NM_019988.5 | NP_064372.2 | 4 | 391 | |||
GenBank mRNA | AK002751.1 | - | 4 | 357 | ||
AK038515.1 | - | 3 | 889 | |||
AK040100.1 | - | 4 | 392 | |||
AK048102.1 | - | 4 | 355 | |||
AK077680.1 | - | 4 | 362 | |||
AK083731.1 | - | 4 | 332 | |||
AK088951.1 | - | 4 | 365 | |||
BC015279.1 | - | 4 | 350 | 4222934 | ||
BM947104.1 | - | 2 | 162 | |||
CF750475.1 | - | 4 | 360 |