Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 7, 8 | 762 | ||
GenBank mRNA | AB476989.1 | - | 7, 8 | 1236 | ||
AB476990.1 | - | 7, 8 | 1122 | |||
AK132271.1 | - | 7, 8 | 785 | |||
BC018390.1 | - | 7, 8 | 762 | 4159877 |