Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 8, 9 | 874 | ||
GenBank mRNA | AB476989.1 | - | 8, 9 | 1348 | ||
AB476990.1 | - | 8, 9 | 1234 | |||
AK132271.1 | - | 8, 9 | 897 | |||
BC018390.1 | - | 8, 9 | 874 | 4159877 | ||
S56368.1 | - | 1, 2 | 102 | |||
U24493.1 | - | 8, 9 | 861 |