Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019989.2 | NP_064373.1 | 2 | 316 | ||
GenBank mRNA | AJ272504.1 | - | 2 | 186 | ||
AK049929.1 | - | 2 | 208 | |||
AK146588.1 | - | 2 | 224 | |||
AK150604.1 | - | 2 | 224 | |||
AK151095.1 | - | 2 | 226 | |||
AK151501.1 | - | 2 | 186 | |||
AK151871.1 | - | 2 | 228 | |||
AK165995.1 | - | 2 | 226 | |||
AK218315.1 | - | 2 | 278 | |||
BC034161.1 | - | 2 | 270 | 4954986 | ||
BC043055.1 | - | 2 | 316 | 6418767 |