Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019977.2 | NP_064361.2 | 7, 8 | 621 | ||
GenBank mRNA | AF197127.1 | - | 7, 8 | 619 | ||
AK002406.1 | - | 7, 8 | 634 | |||
AY738257.1 | - | 6, 7 | 591 | |||
BC013543.1 | - | 7, 8 | 621 | 4242159 |