Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005563.3 | NP_005554.1 | 5 | 575 | ||
NM_203399.1 | NP_981944.1 | 5 | 551 | |||
NM_203401.1 | NP_981946.1 | 5 | 763 | |||
GenBank mRNA | AB209282.1 | - | 4 | 2437 | ||
AB451319.1 | - | 3 | 399 | |||
AB451448.1 | - | 3 | 399 | |||
AK054594.1 | - | 6 | 471 | |||
AK311801.1 | - | 5 | 492 | |||
AV714035.1 | - | 6 | 520 | |||
BC014353.1 | - | 5 | 544 | 3683516 | ||
BC082228.1 | - | 4 | 473 | 2822803 | ||
BF686314.1 | - | 603 | ||||
BG030679.1 | - | 5 | 546 | |||
BM563894.1 | - | 5 | 596 | |||
BX647885.1 | - | 4 | 2451 | |||
J04991.1 | - | 5 | 502 | |||
X53305.1 | - | 5 | 480 | |||
Z11566.1 | - | 5 | 490 |