Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290315.1 | NP_001277244.1 | 20 | 3471 | ||
NM_020043.3 | NP_064427.2 | 20 | 3468 | |||
GenBank mRNA | AB052620.1 | - | 20 | 3468 | ||
AB052621.1 | - | 20 | 3471 | |||
AF176694.1 | - | 20 | 3419 | |||
AK122535.1 | - | 3547 | ||||
BC126880.1 | - | 4 | 386 | |||
CA317106.1 | - | 3 | 411 |