Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204129.1 | NP_001191058.1 | 3 | 287 | ||
NM_001204130.1 | NP_001191059.1 | 3 | 199 | |||
NM_019959.3 | NP_064343.1 | 3 | 277 | |||
GenBank mRNA | AF192499.1 | - | 3 | 225 | ||
AK005484.1 | - | 3 | 246 | |||
AK145104.1 | - | 3 | 200 | |||
BC037636.1 | - | 2 | 192 | |||
BC049894.1 | - | 3 | 228 | 4018549 | ||
CJ056741.1 | - | 3 | 277 | |||
DQ002394.1 | - | 2 | 171 |