Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019981.2 | NP_064365.1 | 3 | 269 | ||
GenBank mRNA | AB022914.1 | - | 3 | 267 | ||
AK005769.1 | - | 3 | 263 | |||
BC048475.1 | - | 3 | 260 | 6740387 | ||
DQ222038.1 | - | 468 | ||||
DQ222039.1 | - | 406 | ||||
DQ222040.1 | - | 769 | ||||
DQ222041.1 | - | 710 | ||||
DQ222042.1 | - | 3 | 271 |