Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253916.1 | NP_001240845.1 | 8 | 1882 | ||
NM_001253917.1 | NP_001240846.1 | 8 | 1882 | |||
NM_019982.3 | NP_064366.2 | 8 | 1882 | |||
GenBank mRNA | AB206790.1 | - | 8 | 1861 | ||
AK083229.1 | - | 8 | 1846 | |||
AK122401.1 | - | 7 | 996 | |||
BC056471.1 | - | 3 | 291 | |||
BC065117.1 | - | 8 | 1842 | 6840338 | ||
BC065129.1 | - | 8 | 1659 | 30362651 | ||
BC079593.1 | - | 2 | 216 | |||
BC094376.1 | - | 2 | 195 | |||
DQ167195.1 | - | 8 | 1869 |