Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002296.3 | NP_002287.2 | 7 | 1054 | ||
NM_194442.2 | NP_919424.1 | 7 | 1044 | |||
XM_005273125.2 | XP_005273182.1 | 7 | 1010 | |||
XM_011544185.2 | XP_011542487.1 | 7 | 1456 | |||
GenBank mRNA | AB209514.1 | - | 7 | 961 | ||
AK222834.1 | - | 7 | 941 | |||
AK303589.1 | - | 7 | 960 | |||
AK312258.1 | - | 7 | 995 | |||
BC020079.1 | - | 7 | 976 | 3925138 | ||
L25931.1 | - | 7 | 955 |