Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020265.4 | NP_064661.2 | 3 | 1187 | ||
GenBank mRNA | AJ243963.2 | - | 3 | 587 | ||
AK028225.1 | - | 3 | 472 | |||
AK031749.1 | - | 3 | 1188 | |||
BC096448.1 | - | 3 | 1178 | 6389874 |