Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000229.1 | NP_000220.1 | 2 | 277 | ||
GenBank mRNA | BC014781.1 | - | 2 | 267 | 4155094 | |
BT009748.1 | - | 2 | 266 | |||
M12625.1 | - | 2 | 533 | |||
M26268.1 | - | 2 | 219 | |||
X06537.1 | - | 2 | 369 |